3-(3-Fluoro-4-methylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166291-5G 166291-1G 166291-500MG
77627-846EA 1484.23 USD
77627-846 77644-792 77648-848
3-(3-Fluoro-4-methylphenyl)-3-oxetanol ≥95%
3-(3-Fluoro-4-methylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 182.19 g/mol
MW: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34851838
CAS Number: 2275624-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR