3-(2-Fluoro-4-methylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166290-1G 166290-500MG 166290-5G
77636-820EA 622 USD
77636-820 77652-388 77653-756
3-(2-Fluoro-4-methylphenyl)-3-oxetanol ≥95%
3-(2-Fluoro-4-methylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₁₀H₁₁FO₂
Storage Temperature: Ambient
MDL Number: MFCD34851837
CAS Number: 2285267-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR