3-[4-(Trimethylsilyl)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166287-1G 166287-5G 166287-500MG
77636-818EA 633 USD
77636-818 77653-754 77644-790
3-[4-(Trimethylsilyl)phenyl]-3-oxetanol ≥95%
3-[4-(Trimethylsilyl)phenyl]-3-oxetanol

Precaución: For Research Use Only

Formula: C₁₂H₁₈O₂Si
Storage Temperature: Ambient
MDL Number: MFCD34851834
CAS Number: 2289321-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR