3-(5-Methoxy-2-methylphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166286-1G 166286-5G 166286-500MG
77636-816EA 507 USD
77636-816 77653-752 77648-846
3-(5-Methoxy-2-methylphenyl)-3-oxetanol ≥95%
3-(5-Methoxy-2-methylphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 194.23 g/mol
MW: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34851833
CAS Number: 2295115-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR