3-[4-(1,1-Dimethylethyl)phenyl]-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166282-1G 166282-5G 166282-500MG
77636-814EA 542 USD
77636-814 77657-602 77648-844
3-[4-(1,1-Dimethylethyl)phenyl]-3-oxetanol ≥95%
3-[4-(1,1-Dimethylethyl)phenyl]-3-oxetanol

Precaución: For Research Use Only

Formula: 206.28 g/mol
MW: C₁₃H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34851830
CAS Number: 1401916-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR