3-(2,4-Dichlorophenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166276-1G 166276-5G 166276-500MG
77636-812EA 507 USD
77636-812 77653-750 77644-788
3-(2,4-Dichlorophenyl)-3-oxetanol ≥95%
3-(2,4-Dichlorophenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₉H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34851825
CAS Number: 1510523-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR