3-(4-Chloro-3-fluorophenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166272-1G 166272-5G 166272-500MG
77636-810EA 507 USD
77636-810 77653-748 77644-786
3-(4-Chloro-3-fluorophenyl)-3-oxetanol ≥95%
3-(4-Chloro-3-fluorophenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD34851822
CAS Number: 1820670-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR