3-(4-Phenoxyphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166270-1G 166270-5G 166270-500MG
77636-808EA 580 USD
77636-808 77653-746 77648-842
3-(4-Phenoxyphenyl)-3-oxetanol ≥95%
3-(4-Phenoxyphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₁₅H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD34851820
CAS Number: 1893871-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR