3-(3,4-Dichlorophenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166257-5G 166257-1G 166257-500MG
77657-600EA 1938 USD
77657-600 77640-808 77644-784
3-(3,4-Dichlorophenyl)-3-oxetanol ≥95%
3-(3,4-Dichlorophenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 219.06 g/mol
MW: C₉H₈Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD34851809
CAS Number: 1820670-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR