3-(2-Fluorophenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166254-500MG 166254-5G 166254-1G
77652-386EA 376 USD
77652-386 77653-744 77640-806
3-(2-Fluorophenyl)-3-oxetanol ≥95%
3-(2-Fluorophenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD33548973
CAS Number: 1904227-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR