3-(3,4-Dimethoxyphenyl)-3-oxetanol ≥97%

Supplier: AOBChem USA

Warning

166248-1G 166248-5G 166248-500MG
77636-804EA 559 USD
77636-804 77657-598 77644-780
3-(3,4-Dimethoxyphenyl)-3-oxetanol ≥97%
3-(3,4-Dimethoxyphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 210.23 g/mol
MW: C₁₁H₁₄O₄
Storage Temperature: Ambient
MDL Number: MFCD34851801
CAS Number: 1523449-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR