3-(3-Methoxyphenyl)-3-oxetanol ≥95%

Supplier: AOBChem USA

Warning

166243-5G 166243-1G 166243-500MG
77657-596EA 1938 USD
77657-596 77640-802 77644-774
3-(3-Methoxyphenyl)-3-oxetanol ≥95%
3-(3-Methoxyphenyl)-3-oxetanol

Precaución: For Research Use Only

Formula: 180.2 g/mol
MW: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD34851799
CAS Number: 1521254-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR