3-(4-(Dimethylamino)phenyl)oxetan-3-ol ≥95%

Supplier: AOBChem USA

Warning

166190-1G 166190-5G 166190-500MG
77636-798EA 507 USD
77636-798 77653-738 77648-838
3-(4-(Dimethylamino)phenyl)oxetan-3-ol ≥95%
3-(4-(Dimethylamino)phenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD34851788
CAS Number: 1903713-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR