3-(4-Fluorophenyl)oxetan-3-ol ≥97%

Supplier: AOBChem USA

Warning

166181-5G 166181-25G 166181-1G 166181-500MG
77631-740EA 1323 USD
77631-740 77657-594 77640-800 77644-772
3-(4-Fluorophenyl)oxetan-3-ol ≥97%
3-(4-Fluorophenyl)oxetan-3-ol

Precaución: For Research Use Only

Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD22370092
CAS Number: 1379812-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR