(3-Chloro-2-fluorophenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

165933-5G 165933-25G 165933-1G 165933-500MG
77631-738EA 1441 USD
77631-738 77653-736 77644-768 77644-770
(3-Chloro-2-fluorophenyl)(ethyl)sulfane ≥95%
(3-Chloro-2-fluorophenyl)(ethyl)sulfane

Precaución: For Research Use Only

Formula: 190.666 g/mol
MW: C₈H₈ClFS
Storage Temperature: Ambient
MDL Number: MFCD22483672
CAS Number: 1824575-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR