2,5-Dimethoxy-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

16520-5G 16520-1G 16520-25G 16520-500MG
77634-126EA 507 USD
77634-126 77636-792 77653-728 77644-764
2,5-Dimethoxy-4-methylphenylboronic acid ≥97%
2,5-Dimethoxy-4-methylphenylboronic acid

Precaución: For Research Use Only

Formula: C₉H₁₃BO₄
Storage Temperature: Ambient
MDL Number: MFCD21602041
CAS Number: 956429-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR