1-Bromo-2-chloro-5-methoxy-4-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

164813-5G 164813-1G 164813-25G 164813-500MG
77631-734EA 1207 USD
77631-734 77636-788 77657-584 77644-760
1-Bromo-2-chloro-5-methoxy-4-methylbenzene ≥95%
1-Bromo-2-chloro-5-methoxy-4-methylbenzene

Precaución: For Research Use Only

Formula: 235.506 g/mol
MW: C₈H₈BrClO
Storage Temperature: Ambient
MDL Number: MFCD28534337
CAS Number: 1782541-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR