Methyl 2-cyclopropylbenzoate ≥97%

Supplier: AOBChem USA

Warning

164676-5G 164676-25G 164676-1G 164676-500MG
77634-122EA 314 USD
77634-122 77653-726 77640-794 77644-758
Methyl 2-cyclopropylbenzoate ≥97%
Methyl 2-cyclopropylbenzoate

Precaución: For Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06802668
CAS Number: 148438-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR