1-(4-Cyclopentylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

164674-1G 164674-500MG 164674-5G
77636-786EA 668 USD
77636-786 77652-382 77653-724
1-(4-Cyclopentylphenyl)ethanone ≥95%
1-(4-Cyclopentylphenyl)ethanone

Precaución: For Research Use Only

Formula: 188.2655 g/mol
MW: C₁₃H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD01095858
CAS Number: 85689-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR