3-(3-Bromophenyl)pentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

164661-5G 164661-1G 164661-25G 164661-500MG
77627-836EA 1777.78 USD
77627-836 77636-784 77653-722 77644-756
3-(3-Bromophenyl)pentan-3-ol ≥97%
3-(3-Bromophenyl)pentan-3-ol

Precaución: For Research Use Only

Formula: 243.1402 g/mol
MW: C₁₁H₁₅BrO
Storage Temperature: Ambient
MDL Number: MFCD28535263
CAS Number: 917024-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR