3-(4-Fluorophenyl)pentan-3-ol ≥97%

Supplier: AOBChem USA

Warning

164657-5G 164657-1G 164657-25G 164657-500MG
77631-732EA 904 USD
77631-732 77636-782 77657-582 77644-754
3-(4-Fluorophenyl)pentan-3-ol ≥97%
3-(4-Fluorophenyl)pentan-3-ol

Precaución: For Research Use Only

Formula: C₁₁H₁₅FO
Storage Temperature: Ambient
MDL Number: MFCD12152944
CAS Number: 76481-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR