1-Bromo-3-(sec-butyl)benzene ≥95%

Supplier: AOBChem USA

Warning

164626-1G 164626-5G 164626-500MG
77636-778EA 532 USD
77636-778 77657-578 77644-752
1-Bromo-3-(sec-butyl)benzene ≥95%
1-Bromo-3-(sec-butyl)benzene

Precaución: For Research Use Only

Formula: C₁₀H₁₃Br
Storage Temperature: Ambient
MDL Number: MFCD26405542
CAS Number: 408307-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR