(±)-1-Phenyl-1-pentanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Fenipentol

164582-5G 164582-25G
77634-120EA 62 USD
77634-120 77653-720
(±)-1-Phenyl-1-pentanol ≥97%
(±)-1-Phenyl-1-pentanol

Precaución: For Research Use Only

Formula: C₆H₅CH(OH)(CH₂)₃CH₃
Storage Temperature: Ambient
MDL Number: MFCD00021935
CAS Number: 583-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR