2-Chloro-4-fluoro-5-methoxyphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-Chloro-4-fluoro-5-methoxyphenyl)boronic acid

16420-5G 16420-25G 16420-1G 16420-500MG
77634-116EA 383 USD
77634-116 77653-716 77640-790 77644-746
2-Chloro-4-fluoro-5-methoxyphenylboronic acid ≥95%
2-Chloro-4-fluoro-5-methoxyphenylboronic acid

Precaución: For Research Use Only

Formula: C₇H₇BClFO₃
MW: 204.391 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998845
CAS Number: 1256355-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR