1-Bromo-4-(phenylmethoxy)-naphthalene ≥97%

Supplier: AOBChem USA

Warning

16379-5G 16379-1G 16379-25G 16379-500MG
77634-114EA 325 USD
77634-114 77636-772 77653-712 77648-830
1-Bromo-4-(phenylmethoxy)-naphthalene ≥97%
1-Bromo-4-(phenylmethoxy)-naphthalene

Precaución: For Research Use Only

Formula: 313.189 g/mol
MW: C₁₇H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD27756646
CAS Number: 138865-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR