1-(3-Bromo-5-ethyl-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
Supplier: AOBChem USA
|
Warning
|
163785-5G
163785-25G
163785-1G
163785-500MG
77627-832EA
1484.23
USD
77627-832
77657-574
77640-788
77644-744
1-(3-Bromo-5-ethyl-2-fluorophenyl)-2,2,2-trifluoroethanone ≥97%
1-(3-Bromo-5-ethyl-2-fluorophenyl)-2,2,2-trifluoroethanone
Precaución: For Research Use Only
Formula:
C₁₀H₇BrF₄O Storage Temperature: Ambient |
MDL Number:
MFCD34850222 CAS Number: 2921683-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...