2,6-Dichloro-3-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16329-5G 16329-25G 16329-1G 16329-500MG
77634-112EA 564 USD
77634-112 77657-566 77640-784 77648-824
2,6-Dichloro-3-methoxybenzaldehyde ≥97%
2,6-Dichloro-3-methoxybenzaldehyde

Precaución: For Research Use Only

Formula: 205.038 g/mol
MW: C₈H₆Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26793804
CAS Number: 6505-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR