2,6-Dichloro-3,5-dimethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

16310-5G 16310-1G 16310-500MG
77627-828EA 1777.78 USD
77627-828 77636-766 77644-738
2,6-Dichloro-3,5-dimethoxybenzaldehyde ≥95%
2,6-Dichloro-3,5-dimethoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₈Cl₂O₃
MW: 235.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD25970240
CAS Number: 287175-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR