(4-Methyl-1,3-phenylene)bis(methylsulfane) ≥95%

Supplier: AOBChem USA

Warning

162912-5G 162912-1G 162912-25G 162912-500MG
77627-826EA 1777.78 USD
77627-826 77636-764 77657-564 77648-822
(4-Methyl-1,3-phenylene)bis(methylsulfane) ≥95%
(4-Methyl-1,3-phenylene)bis(methylsulfane)

Precaución: For Research Use Only

Formula: 184.3215 g/mol
MW: C₉H₁₂S₂
Storage Temperature: Ambient
MDL Number: MFCD34849593
CAS Number: 2921850-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR