(3-Bromo-4-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

162905-5G 162905-25G 162905-1G 162905-500MG
77631-724EA 1441 USD
77631-724 77653-710 77640-782 77644-736
(3-Bromo-4-methylphenyl)(methyl)sulfane ≥95%
(3-Bromo-4-methylphenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: C₈H₉BrS
Storage Temperature: Ambient
MDL Number: MFCD15527263
CAS Number: 866997-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR