2-Chloro-3-ethoxy-6-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16280-5G 16280-25G 16280-1G 16280-500MG
77631-722EA 1316 USD
77631-722 77653-708 77640-780 77644-734
2-Chloro-3-ethoxy-6-fluorobenzaldehyde ≥97%
2-Chloro-3-ethoxy-6-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD25960239
CAS Number: 181305-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR