4-Chloro-1-ethoxy-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

16276-5G 16276-25G 16276-1G 16276-500MG
77634-110EA 436 USD
77634-110 77653-706 77640-778 77644-732
4-Chloro-1-ethoxy-2-fluorobenzene ≥97%
4-Chloro-1-ethoxy-2-fluorobenzene

Precaución: For Research Use Only

Formula: C₈H₈ClFO
Storage Temperature: Ambient
MDL Number: MFCD00672982
CAS Number: 289039-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR