6-Chloro-3-ethoxy-2-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16275-5G 16275-25G 16275-1G 16275-500MG
77631-720EA 835 USD
77631-720 77653-704 77640-776 77644-730
6-Chloro-3-ethoxy-2-fluorobenzaldehyde ≥97%
6-Chloro-3-ethoxy-2-fluorobenzaldehyde

Precaución: For Research Use Only

Formula: 202.61 g/mol
MW: C₉H₈ClFO₂
Storage Temperature: Ambient
MDL Number: MFCD25962746
CAS Number: 363620-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR