6-Bromo-2-chloro-3-ethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

16255-5G 16255-500MG 16255-25G 16255-1G
77634-108EA 382 USD
77634-108 77652-376 77657-560 77640-774
6-Bromo-2-chloro-3-ethoxybenzaldehyde ≥95%
6-Bromo-2-chloro-3-ethoxybenzaldehyde

Precaución: For Research Use Only

Formula: 263.516 g/mol
MW: C₉H₈BrClO₂
Storage Temperature: Ambient
MDL Number: MFCD28976862
CAS Number: 704898-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR