5-Bromo-2-(2-methyl-1,3-dioxolan-2-yl)pyridine ≥95%

Supplier: AOBChem USA

Warning

16241-500MG 16241-5G 16241-1G
77652-374EA 419 USD
77652-374 77657-558 77640-772
5-Bromo-2-(2-methyl-1,3-dioxolan-2-yl)pyridine ≥95%
5-Bromo-2-(2-methyl-1,3-dioxolan-2-yl)pyridine

Precaución: For Research Use Only

Formula: 244.085 g/mol
MW: C₉H₁₀BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD14706790
CAS Number: 214701-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR