Ethyl 2-(6-methoxypyridin-3-yl)-2-oxoacetate ≥97%

Supplier: AOBChem USA

Warning

162203-5G 162203-25G 162203-1G 162203-500MG
77634-106EA 522 USD
77634-106 77657-552 77640-768 77644-726
Ethyl 2-(6-methoxypyridin-3-yl)-2-oxoacetate ≥97%
Ethyl 2-(6-methoxypyridin-3-yl)-2-oxoacetate

Precaución: For Research Use Only

Formula: C₁₀H₁₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD21603986
CAS Number: 1280197-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR