Ethyl-2-oxo-2-(pyridin-3-yl)acetate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-oxo-2-(pyridin-3-yl)acetate

162201-5G 162201-500MG 162201-25G 162201-1G
77634-104EA 455 USD
77634-104 77652-372 77653-698 77640-766
Ethyl-2-oxo-2-(pyridin-3-yl)acetate ≥97%
Ethyl-2-oxo-2-(pyridin-3-yl)acetate

Precaución: For Research Use Only

Formula: C₉H₉NO₃
MW: 179.1727 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09842445
CAS Number: 73672-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR