4-Chloro-2-fluoro-3-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

16211-5G 16211-1G 16211-25G 16211-500MG
77634-102EA 288 USD
77634-102 77636-754 77657-550 77644-724
4-Chloro-2-fluoro-3-iodobenzaldehyde ≥97%
4-Chloro-2-fluoro-3-iodobenzaldehyde

Precaución: For Research Use Only

Formula: C₇H₃ClFIO
Storage Temperature: Ambient
MDL Number: MFCD26400343
CAS Number: 909186-22-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR