Methyl 4,5-dichloro-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

161791-1G 161791-500MG 161791-5G
77636-752EA 546 USD
77636-752 77652-370 77657-548
Methyl 4,5-dichloro-2-fluorobenzoate ≥97%
Methyl 4,5-dichloro-2-fluorobenzoate

Precaución: For Research Use Only

Formula: C₈H₅Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD14698031
CAS Number: 1214336-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR