Ethyl(2-fluoro-3-methylphenyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

161546-5G 161546-25G 161546-1G 161546-500MG
77631-384EA 1553.24 USD
77631-384 77635-464 77636-748 77648-810
Ethyl(2-fluoro-3-methylphenyl)sulfane ≥95%
Ethyl(2-fluoro-3-methylphenyl)sulfane

Precaución: For Research Use Only

Formula: C₉H₁₁FS
Storage Temperature: Ambient
MDL Number: MFCD22484212
CAS Number: 252555-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR