(4-Chloro-2-methylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

161543-5G 161543-25G 161543-1G 161543-500MG
77631-708EA 1180 USD
77631-708 77653-696 77640-764 77644-722
(4-Chloro-2-methylphenyl)(ethyl)sulfane ≥95%
(4-Chloro-2-methylphenyl)(ethyl)sulfane

Precaución: For Research Use Only

Formula: C₉H₁₁ClS
Storage Temperature: Ambient
MDL Number: MFCD11617740
CAS Number: 1314923-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR