(3-Ethyl-4-fluoro-5-methylphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

161216-5G 161216-1G 161216-500MG 161216-25G
77631-706EA 1441 USD
77631-706 77636-746 77652-368 77653-694
(3-Ethyl-4-fluoro-5-methylphenyl)(methyl)sulfane ≥95%
(3-Ethyl-4-fluoro-5-methylphenyl)(methyl)sulfane

Precaución: For Research Use Only

Formula: 184.274 g/mol
MW: C₁₀H₁₃FS
Storage Temperature: Ambient
MDL Number: MFCD34848500
CAS Number: 2710913-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR