Cyclopentylbenzene ≥97%

Supplier: AOBChem USA

Warning

161024-5G 161024-25G 161024-1G 161024-500MG
77634-100EA 511 USD
77634-100 77653-692 77640-762 77644-720
Cyclopentylbenzene ≥97%
Cyclopentylbenzene

Precaución: For Research Use Only

Formula: C₁₁H₁₄
Storage Temperature: Ambient
MDL Number: MFCD00019287
CAS Number: 700-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR