(4-Chloro-2-fluoro-5-nitrophenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

161003-1G 161003-500MG
77634-098EA 1483 USD
77634-098 77648-808
(4-Chloro-2-fluoro-5-nitrophenyl)boronic acid ≥97%
(4-Chloro-2-fluoro-5-nitrophenyl)boronic acid

Precaución: For Research Use Only

Formula: C₆H₄BClFNO₄
Storage Temperature: Ambient
CAS Number: 325786-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR