(7-Chloro-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

160950-5G 160950-25G 160950-1G 160950-500MG
77631-382EA 1777.78 USD
77631-382 77653-690 77640-760 77648-806
(7-Chloro-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)boronic acid ≥97%
(7-Chloro-2,3-dihydrobenzo[b][1,4]dioxin-6-yl)boronic acid

Precaución: For Research Use Only

Formula: 214.411 g/mol
MW: C₈H₈BClO₄
Storage Temperature: Ambient
MDL Number: MFCD34848330
CAS Number: 2770006-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR