3-Fluoro-2,4-dimethoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (3-Fluoro-2,4-dimethoxyphenyl)boronic acid

16083-5G 16083-25G
77634-096EA 84 USD
77634-096 77657-544
3-Fluoro-2,4-dimethoxyphenylboronic acid ≥97%
3-Fluoro-2,4-dimethoxyphenylboronic acid

Precaución: For Research Use Only

Formula: C₈H₁₀BFO₄
MW: 199.972 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23381042
CAS Number: 949146-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR