2,6-Difluoro-4-propylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

16066-5G 16066-25G 16066-1G 16066-500MG
77634-094EA 539 USD
77634-094 77653-688 77640-758 77644-718
2,6-Difluoro-4-propylphenylboronic acid ≥95%
2,6-Difluoro-4-propylphenylboronic acid

Precaución: For Research Use Only

Formula: C₉H₁₁BF₂O₂
MW: 199.99 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21337851
CAS Number: 1010110-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR