(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

160618-5G 160618-1G 160618-25G 160618-500MG
77631-704EA 1441 USD
77631-704 77636-744 77657-542 77648-804
(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)trimethylsilane ≥95%
(2-Fluoro-3-(methylthio)-5-(trifluoromethyl)phenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₁H₁₄F₄SSi
Storage Temperature: Ambient
MDL Number: MFCD35166598
CAS Number: 3002486-44-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR