(2-Fluoro-3-methyl-5-(methylthio)phenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

160608-5G 160608-1G 160608-25G 160608-500MG
77631-702EA 1441 USD
77631-702 77636-742 77653-686 77648-802
(2-Fluoro-3-methyl-5-(methylthio)phenyl)trimethylsilane ≥97%
(2-Fluoro-3-methyl-5-(methylthio)phenyl)trimethylsilane

Precaución: For Research Use Only

Formula: C₁₁H₁₇FSSi
Storage Temperature: Ambient
MDL Number: MFCD34848101
CAS Number: 2969372-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR