1-(2,4-Difluoro-3-methylphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

160539-5G 160539-25G 160539-1G 160539-500MG
77631-700EA 1206 USD
77631-700 77657-540 77640-756 77648-800
1-(2,4-Difluoro-3-methylphenyl)ethanone ≥95%
1-(2,4-Difluoro-3-methylphenyl)ethanone

Precaución: For Research Use Only

Formula: C₉H₈F₂O
Storage Temperature: Ambient
MDL Number: MFCD24711330
CAS Number: 112857-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR